Assay Details
Gene Symbol: IFNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081949.1 | NP_001075418.1 | 3 - 4 | 428 | 76 | |
GenBank mRNA | D28520.1 | - | 3 - 4 | 364 | 76 | |
EU000433.1 | - | 3 - 4 | 364 | 76 | ||
JL628261.1 | - | 1 - 2 | 190 | 76 | ||
KJ814948.1 | - | 3 - 4 | 304 | 76 | ||
U04050.1 | - | 3 - 4 | 428 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
