Assay Details
Gene Symbol: IL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082490.1 | NP_001075959.1 | 1 - 2 | 165 | 78 | |
XM_023640225.1 | XP_023495993.1 | 7495 | 78 | |||
GenBank mRNA | EU438771.1 | - | 1 - 2 | 165 | 78 | |
JQ432545.1 | - | 1 - 2 | 165 | 78 | ||
U38200.1 | - | 1 - 2 | 165 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
