Assay Details
Gene Symbol: IL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082496.2 | NP_001075965.2 | 2 - 3 | 204 | 83 | |
GenBank mRNA | AF005227.1 | - | 1 - 2 | 204 | 83 | |
AF041975.1 | - | 1 - 2 | 204 | 83 | ||
EU438770.1 | - | 1 - 2 | 198 | 83 | ||
KX345376.1 | - | 1 - 2 | 123 | 83 | ||
U64794.1 | - | 2 - 3 | 204 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
