Assay Details
Gene Symbol: IL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082499.1 | NP_001075968.1 | 2 - 3 | 184 | 99 | |
GenBank mRNA | U91947.1 | - | 2 - 3 | 184 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
