Assay Details
Gene Symbol: B2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082502.3 | NP_001075971.1 | 1 - 2 | 94 | 70 | |
XM_005602595.3 | XP_005602652.1 | 146 | 70 | |||
GenBank mRNA | CD464355.1 | - | 1 - 2 | 101 | 70 | |
DN506758.1 | - | 1 - 2 | 67 | 70 | ||
JL616344.2 | - | 1 - 2 | 95 | 70 | ||
X69083.1 | - | 72 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
