Assay Details
Gene Symbol: IL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082512.1 | NP_001075981.1 | 3 - 4 | 226 | 117 | |
XM_005611426.3 | XP_005611483.1 | 503 | 117 | |||
XM_005611427.3 | XP_005611484.1 | 497 | 117 | |||
GenBank mRNA | EU438772.1 | - | 3 - 4 | 238 | 117 | |
JQ432547.1 | - | 1 - 2 | 118 | 117 | ||
Y11131.1 | - | 226 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
