Assay Details
Gene Symbol: IL18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082512.1 | NP_001075981.1 | 4 - 5 | 364 | 143 | |
XM_005611426.3 | XP_005611483.1 | 641 | 143 | |||
XM_005611427.3 | XP_005611484.1 | 635 | 143 | |||
GenBank mRNA | EU438772.1 | - | 4 - 5 | 376 | 143 | |
JL640763.1 | - | 2 - 3 | 274 | 143 | ||
JQ432547.1 | - | 2 - 3 | 256 | 143 | ||
Y11131.1 | - | 364 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
