Assay Details
Gene Symbol: DNASE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082514.2 | NP_001075983.1 | 2 - 3 | 320 | 61 | |
XM_023655008.1 | XP_023510776.1 | 3386 | 61 | |||
GenBank mRNA | AB162819.1 | - | 2 - 3 | 320 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
