Assay Details
Gene Symbol: TIMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082515.1 | NP_001075984.1 | 4 - 5 | 473 | 93 | |
XM_023633181.1 | XP_023488949.1 | 622 | 93 | |||
GenBank mRNA | U95039.1 | - | 4 - 5 | 473 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
