Assay Details
Gene Symbol: CYP17A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082523.1 | NP_001075992.1 | 6 - 7 | 1174 | 73 | |
GenBank mRNA | D30688.1 | - | 6 - 7 | 1174 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
