Assay Details
Gene Symbol: HSD17B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083598.2 | NP_001077067.1 | 2 - 3 | 294 | 58 | |
XM_023651598.1 | XP_023507366.1 | 356 | 58 | |||
GenBank mRNA | DQ418451.1 | - | 2 - 3 | 294 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
