Assay Details
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085433.2 | NP_001078902.1 | 1 - 2 | 171 | 88 | |
GenBank mRNA | EU438768.1 | - | 1 - 2 | 169 | 88 | |
JQ432544.1 | - | 1 - 2 | 140 | 88 | ||
X69393.1 | - | 171 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
