Assay Details
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085433.2 | NP_001078902.1 | 3 - 4 | 368 | 141 | |
GenBank mRNA | EU438768.1 | - | 3 - 4 | 366 | 141 | |
JQ432544.1 | - | 3 - 4 | 337 | 141 | ||
X69393.1 | - | 368 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
