Assay Details
Gene Symbol: RNASEL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089586.1 | NP_001083055.1 | 3 - 4 | 1762 | 78 | |
GenBank mRNA | DQ497159.1 | - | 3 - 4 | 1762 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
