Assay Details
Gene Symbol: CA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099760.1 | NP_001093230.1 | 1 - 2 | 94 | 87 | |
XM_005602598.3 | XP_005602655.2 | 391 | 87 | |||
GenBank mRNA | EF397505.1 | - | 1 - 2 | 94 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
