Assay Details
Gene Symbol: CA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099760.1 | NP_001093230.1 | 2 - 3 | 119 | 116 | |
XM_005602598.3 | XP_005602655.2 | 416 | 116 | |||
GenBank mRNA | EF397505.1 | - | 2 - 3 | 119 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
