Assay Details
Gene Symbol: CD68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099762.1 | NP_001093232.1 | 2 - 3 | 530 | 147 | |
GenBank mRNA | AY373259.2 | - | 2 - 3 | 530 | 147 | |
AY499346.1 | - | 2 - 3 | 489 | 147 | ||
JL615906.1 | - | 2 - 3 | 555 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
