Assay Details
Gene Symbol: CD68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099762.1 | NP_001093232.1 | 4 - 5 | 719 | 70 | |
GenBank mRNA | AY373259.2 | - | 4 - 5 | 719 | 70 | |
AY499346.1 | - | 4 - 5 | 678 | 70 | ||
JL615906.1 | - | 4 - 5 | 744 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
