Assay Details
Gene Symbol: PLA2G2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100113.3 | NP_001093583.1 | 2 - 3 | 242 | 115 | |
XM_005607005.3 | XP_005607062.2 | 540 | 115 | |||
XM_023626373.1 | XP_023482141.1 | 608 | 115 | |||
XM_023626382.1 | XP_023482150.1 | 630 | 115 | |||
XM_023626385.1 | XP_023482153.1 | 320 | 115 | |||
GenBank mRNA | EF428565.1 | - | 2 - 3 | 243 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
