Assay Details
Gene Symbol: VCAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101650.1 | NP_001095120.1 | 6 - 7 | 1590 | 64 | |
XM_005610265.3 | XP_005610322.1 | 1964 | 64 | |||
GenBank mRNA | EU057895.1 | - | 6 - 7 | 1590 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
