Assay Details
Gene Symbol: RSPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109681.1 | NP_001103151.1 | 3 - 4 | 427 | 96 | |
GenBank mRNA | EU029661.1 | - | 3 - 4 | 427 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
