Assay Details
Gene Symbol: IFI27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114146.1 | NP_001107618.1 | 3 - 4 | 246 | 89 | |
XM_005605175.3 | XP_005605232.1 | 467 | 89 | |||
GenBank mRNA | BM735054.1 | - | 3 - 4 | 246 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
