Assay Details
Gene Symbol: CXCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114940.1 | NP_001108412.1 | 1 - 2 | 53 | 79 | |
GenBank mRNA | EU438777.1 | - | 1 - 2 | 53 | 79 | |
HQ890261.1 | - | 1 - 2 | 119 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
