Assay Details
Gene Symbol: HSP90AA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163955.1 | NP_001157427.1 | 6 - 7 | 1342 | 116 | |
XM_023627692.1 | XP_023483460.1 | 1520 | 116 | |||
GenBank mRNA | AB043677.1 | - | 6 - 7 | 1309 | 116 | |
HQ890110.1 | - | 3 - 4 | 667 | 116 | ||
JL617688.1 | - | 4 - 5 | 794 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
