Assay Details
Gene Symbol: HSP90AA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163955.1 | NP_001157427.1 | 7 - 8 | 1494 | 83 | |
XM_023627692.1 | XP_023483460.1 | 1672 | 83 | |||
GenBank mRNA | AB043677.1 | - | 7 - 8 | 1461 | 83 | |
HQ890110.1 | - | 4 - 5 | 819 | 83 | ||
JL617688.1 | - | 5 - 6 | 946 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
