Assay Details
Gene Symbol: IGFBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491171.5 | XP_001491221.3 | 740 | 85 | ||
GenBank mRNA | AB072934.1 | - | 2 - 3 | 225 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
