Assay Details
Gene Symbol: MYH4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_015344.1 | 1 - 2 | 175 | 73 | ||
GenBank mRNA | AB126008.1 | - | 1 - 2 | 175 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
