Assay Details
Gene Symbol: PKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143794.2 | NP_001137266.1 | 5 - 6 | 722 | 82 | |
NM_001159690.2 | NP_001153162.1 | 5 - 6 | 722 | 82 | ||
XM_023641094.1 | XP_023496862.1 | 728 | 82 | |||
XM_023641126.1 | XP_023496894.1 | 731 | 82 | |||
GenBank mRNA | AB355734.1 | - | 5 - 6 | 666 | 82 | |
AB355735.1 | - | 5 - 6 | 722 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
