Assay Details
Gene Symbol: KITLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163962.2 | NP_001157434.1 | 7 - 8 | 901 | 79 | |
XM_005606468.3 | XP_005606525.1 | 1158 | 79 | |||
GenBank mRNA | AF053498.1 | - | 6 - 7 | 685 | 79 | |
AF401625.1 | - | 6 - 7 | 710 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
