Assay Details
Gene Symbol: ATP1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640511.1 | XP_023496279.1 | 989 | 65 | ||
GenBank mRNA | AJ539224.1 | - | 2 - 3 | 318 | 65 | |
EU423855.1 | - | 2 - 3 | 297 | 65 | ||
JL616040.1 | - | 3 - 4 | 479 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
