Assay Details
Gene Symbol: AQP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917787.5 | XP_001917822.2 | 287 | 73 | ||
GenBank mRNA | AM182511.1 | - | 1 - 2 | 53 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
