Assay Details
Gene Symbol: LDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145111.1 | NP_001138583.1 | 2 - 3 | 231 | 57 | |
XM_014740462.2 | XP_014595948.2 | 160 | 57 | |||
GenBank mRNA | AB472336.1 | - | 2 - 3 | 224 | 57 | |
AB477534.1 | - | 2 - 3 | 231 | 57 | ||
AY246702.1 | - | 2 - 3 | 183 | 57 | ||
JL616405.2 | - | 2 - 3 | 224 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
