Assay Details
Gene Symbol: DET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005602804.3 | XP_005602861.1 | 575 | 97 | ||
XM_005602806.3 | XP_005602863.1 | 745 | 97 | |||
XM_005602807.3 | XP_005602864.1 | 676 | 97 | |||
XM_023650329.1 | XP_023506097.1 | 1006 | 97 | |||
XM_023650337.1 | XP_023506105.1 | 676 | 97 | |||
GenBank mRNA | AY246761.1 | - | 1 | 255 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
