Assay Details
Gene Symbol: HSPA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163884.2 | NP_001157356.2 | 10 - 11 | 1277 | 91 | |
GenBank mRNA | AY348323.1 | - | 1 - 2 | 86 | 91 | |
HQ889946.1 | - | 3 - 4 | 404 | 91 | ||
JL640522.1 | - | 4 - 5 | 573 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
