Assay Details
Gene Symbol: HPRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023634464.1 | XP_023490232.1 | 275 | 84 | ||
GenBank mRNA | AY372182.1 | - | 1 - 2 | 111 | 84 | |
JL616035.2 | - | 2 - 3 | 145 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
