Assay Details
Gene Symbol: PITPNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163878.1 | NP_001157350.1 | 5 - 6 | 1062 | 65 | |
XM_005597019.3 | XP_005597076.1 | 1253 | 65 | |||
XM_023651592.1 | XP_023507360.1 | 1314 | 65 | |||
XM_023651593.1 | XP_023507361.1 | 461 | 65 | |||
XM_023651594.1 | XP_023507362.1 | 389 | 65 | |||
XM_023651596.1 | XP_023507364.1 | 1313 | 65 | |||
GenBank mRNA | AY500592.1 | - | 4 - 5 | 246 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
