Assay Details
Gene Symbol: RBM8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163973.1 | NP_001157445.1 | 4 - 5 | 401 | 134 | |
XM_005609925.3 | XP_005609982.1 | 426 | 134 | |||
GenBank mRNA | AY647161.1 | - | 3 - 4 | 252 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
