Assay Details
Gene Symbol: EDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163964.1 | NP_001157436.1 | 2 - 3 | 236 | 78 | |
GenBank mRNA | AY730629.1 | - | 1 - 2 | 82 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
