Assay Details
Gene Symbol: EDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163964.1 | NP_001157436.1 | 3 - 4 | 391 | 96 | |
GenBank mRNA | AY730629.1 | - | 2 - 3 | 237 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
