Assay Details
Gene Symbol: IGFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001308602.2 | NP_001295531.1 | 3 - 4 | 789 | 78 | |
GenBank mRNA | AY752930.1 | - | 2 - 3 | 278 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
