Assay Details
Gene Symbol: IGFBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023642295.1 | XP_023498063.1 | 908 | 54 | ||
GenBank mRNA | AY752931.1 | - | 2 - 3 | 288 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
