Assay Details
Gene Symbol: GPX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504880.3 | XP_001504930.1 | 471 | 139 | ||
GenBank mRNA | DQ058077.1 | - | 1 - 2 | 29 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
