Assay Details
Gene Symbol: MDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195526.2 | NP_001182455.1 | 4 - 5 | 539 | 58 | |
GenBank mRNA | DQ402962.1 | - | 3 - 4 | 276 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
