Assay Details
Gene Symbol: SDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163823.2 | NP_001157295.1 | 5 - 6 | 545 | 71 | |
GenBank mRNA | DQ403012.1 | - | 5 - 6 | 490 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
