Assay Details
Gene Symbol: IDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195527.1 | NP_001182456.1 | 5 - 6 | 855 | 65 | |
XM_005601394.3 | XP_005601451.1 | 1084 | 65 | |||
GenBank mRNA | DQ403087.1 | - | 4 - 5 | 714 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
