Assay Details
Gene Symbol: CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195515.1 | NP_001182444.1 | 10 - 11 | 1391 | 83 | |
GenBank mRNA | DQ403137.1 | - | 4 - 5 | 566 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
