Assay Details
Gene Symbol: SLCO1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309511.1 | NP_001296440.1 | 3 - 4 | 336 | 89 | |
XM_014740471.2 | XP_014595957.1 | 733 | 89 | |||
XM_014740474.2 | XP_014595960.1 | 592 | 89 | |||
XM_023642170.1 | XP_023497938.1 | 1185 | 89 | |||
XM_023642171.1 | XP_023497939.1 | 1461 | 89 | |||
XM_023642172.1 | XP_023497940.1 | 915 | 89 | |||
XM_023642173.1 | XP_023497941.1 | 1149 | 89 | |||
XM_023642174.1 | XP_023497942.1 | 1181 | 89 | |||
XM_023642175.1 | XP_023497943.1 | 1186 | 89 | |||
XM_023642176.1 | XP_023497944.1 | 1186 | 89 | |||
XM_023642177.1 | XP_023497945.1 | 1045 | 89 | |||
XM_023642179.1 | XP_023497947.1 | 451 | 89 | |||
GenBank mRNA | DQ409214.1 | - | 2 - 3 | 186 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
