Assay Details
Gene Symbol: IL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143791.1 | NP_001137263.1 | 1 - 2 | 128 | 62 | |
XM_023616897.1 | XP_023472665.1 | 1397 | 62 | |||
GenBank mRNA | DQ889711.1 | - | 1 - 2 | 120 | 62 | |
EF645663.1 | - | 1 - 2 | 128 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
