Assay Details
Gene Symbol: TIMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023651899.1 | XP_023507667.1 | 535 | 72 | ||
GenBank mRNA | AJ010315.1 | - | 1 - 2 | 97 | 72 | |
EF077283.1 | - | 2 - 3 | 167 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
