Assay Details
Gene Symbol: NOX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495665.6 | XP_001495715.4 | 2178 | 64 | ||
GenBank mRNA | EF152773.1 | - | 4 - 5 | 619 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
